
cis-13,16-Docosadienoic acid methyl ester

Empirical Formula (Hill Notation):
C23H42O2
CAS Number:
Molecular Weight:
350.58
Products
Product Category
Brand
Biological Source
Formula Weight
Physical Form
Purity
Quality Segment
Functional Group
Showing 1-1 of 1