CEFTIOFUR HYDROCHLORIDE
Synonym(s):
Ceftiofur hydrochloride, (6R,7R)-7-{[(2Z)-2-(2-Amino-4-thiazolyl)-2-(methoxyimino)acetyl]amino}-3-{[(2-furanylcarbonyl)thio]methyl}-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid hydrochloride, 3-[(2-Furylcarbonyl)thio]methyl]-7-[2-(2-amino-4-thiazolyl)-2-(methoxyiminoacetamido)-3-cephem-4-carboxylic acid hydrochloride
Empirical Formula (Hill Notation):
C19H17N5O7S3 · HCl
CAS Number:
Molecular Weight:
560.02
Products
Product Category
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing