Skip to Content
Merck
Bis(trichloromethyl) carbonate purum, ≥99.0% (AT)

Bis(trichloromethyl) carbonate

Bis(trichloromethyl) carbonate purum, ≥99.0% (AT)
Synonym(s):
Triphosgene, tri-Phosgene, Bis(trichloromethyl) carbonate
Linear Formula:
(Cl3CO)2CO
CAS Number:
Molecular Weight:
296.75

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing