
Bismerthiazol

Synonym(s):
N,N′-Methylene-bis(2-amino-5-sulfhydryl-1,3,4-thiadiazole)
Empirical Formula (Hill Notation):
C5H6N6S4
CAS Number:
Molecular Weight:
278.40
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Product Line
Format
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing