![Anti-STAT1 antibody produced in rabbit Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution](/deepweb/assets/sigmaaldrich/product/images/297/487/bca73dda-8f4a-4278-b9c1-e05fd31f92a6/640/bca73dda-8f4a-4278-b9c1-e05fd31f92a6.jpg)
Anti-STAT1 antibody produced in rabbit
![Anti-STAT1 antibody produced in rabbit Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution](/deepweb/assets/sigmaaldrich/product/images/297/487/bca73dda-8f4a-4278-b9c1-e05fd31f92a6/640/bca73dda-8f4a-4278-b9c1-e05fd31f92a6.jpg)
Synonym(s):
Anti-Signal transducer and activator of transcription 1-α/β antibody produced in rabbit, Anti-Transcription factor ISGF-3 components p91/p84 antibody produced in rabbit, Signal transducer and activator of transcription 1-α/β, Transcription factor ISGF-3 components p91/p84
Products
Product Category
Brand
Biological Source
Clonality
Feature
Gene Alias
Physical Form
Product Line
Antibody Form
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique
Showing 1-4 of 4
Compare
Product Number
Description
Pricing