Skip to Content
Merck
Ammonium iron(III) oxalate trihydrate ≥98%

Ammonium iron(III) oxalate trihydrate

Ammonium iron(III) oxalate trihydrate ≥98%
Synonym(s):
Ammonium ferric oxalate, Iron ammonium oxalate
Linear Formula:
(NH4)3[Fe(C2O4)3] · 3H2O
CAS Number:
Molecular Weight:
428.06
EC Number:

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing