
8-(Diethylamino)octyl-3,4,5-trimethoxybenzoate hydrochloride

Synonym(s):
TMB 8 hydrochloride, TMB-8 · HCl
Linear Formula:
(CH3O)3C6H2CO2(CH2)8N(C2H5)2 · HCl
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing