4-(N-Maleimidomethyl)cyclohexanecarboxylic acid N-hydroxysuccinimide ester
Synonym(s):
N-Succinimidyl 4-(maleimidomethyl)cyclohexanecarboxylate, SMCC
Empirical Formula (Hill Notation):
C16H18N2O6
CAS Number:
Molecular Weight:
334.32
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Reagent Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing