
2,4,6-Tribromo-3-hydroxybenzoic acid

Synonym(s):
3-Hydroxy-2,4,6-tribromobenzoic acid
Linear Formula:
HC6Br3(OH)CO2H
CAS Number:
Molecular Weight:
374.81
Products
Product Category
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing