Skip to Content
Merck
All Photos(1)

Key Documents

57277

Supelco

Ferrules/Nuts for Visiprep Large Volume Sampler

color-coded, pk of 4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12000000

feature

color-coded

packaging

pk of 4

technique(s)

solid phase extraction (SPE): suitable

fitting

1/4-28 male UNF fittings

application(s)

food and beverages

General description

Ferrules/Nuts for Visiprep Large Volume Sampler are accessories for use with Visiprep large volume samplers which enable the transfer of low viscosity samples directly from any sample container to conventional solid phase extraction (SPE) tubes on the Visiprep SPE vacuum manifolds.

Legal Information

Visiprep is a trademark of Sigma-Aldrich Co. LLC

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service