Skip to Content
Merck
All Photos(1)

Key Documents

22018-U

Swagelok® Caps

brass, 1/8 in. male

Synonym(s):

Swagelok® 200C,BRASS CAPS 1/8" PK6

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41112500
eCl@ss:
32035302
NACRES:
SB.54

material

brass

packaging

pkg of 6 ea (nuts & ferrules included)

fitting

1/8 in. male

Looking for similar products? Visit Product Comparison Guide

General description

Nuts and ferrules included.

Application

Swagelok® caps maybe used to seal the conditioned sampling tube, during gas chromatographic determination of low concentrations of volatile organic compounds (≤C6) in ambient air using mercuric oxide reduction gas detection (RGD).

Legal Information

Swagelok is a registered trademark of Swagelok Company

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Passive sampling and gas chromatographic determination of low concentrations of reactive hydrocarbons in ambient air with reduction gas detector.
Xu-Liang C and Hewitt N.
Journal of Chromatography A, 648 (1), 191-197 (1993)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service