Skip to Content
Merck
All Photos(1)

Key Documents

SRP4226

Sigma-Aldrich

MCP-2 from mouse

recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC)

Synonym(s):

CCL8, HC14, MCP-2, Monocyte chemoattractant protein 2, Monocyte chemotactic protein 2, Small inducible cytokine A8

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.32

biological source

mouse

recombinant

expressed in E. coli

Assay

≥98% (HPLC)
≥98% (SDS-PAGE)

form

lyophilized

mol wt

~8.5 kDa

packaging

pkg of 10 μg

impurities

endotoxin, tested

NCBI accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... Ccl8(20307)

General description

Macrophage/monocyte chemotactic protein-2 (MCP-2) plays an important role in the inflammatory response of blood monocytes, activated T cells, basophils, NK cells, and immature dendritic cells. Recombinant mouse MCP-2 is an 8.5 kDa protein containing 74 amino acid residues.

Physical form

Sterile filtered and then lyophilized without any additives.

Reconstitution

Centrifuge the vial prior to opening. Avoid freeze-thaw cycles.
Reconstitute in water to a concentration of 0.1-1.0 mg/mL. This solution can then be diluted into other aqueous buffers.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service