Skip to Content
Merck
All Photos(1)

Key Documents

79622-M

Sigma-Aldrich

Phosphoric acid

puriss. p.a., crystallized, ≥99.0% (T)

Synonym(s):

Orthophosphoric acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H3PO4
CAS Number:
Molecular Weight:
98.00
Beilstein:
1921286
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.21

vapor density

3.4 (vs air)

Quality Level

vapor pressure

2.2 mmHg ( 20 °C)
5 mmHg ( 25 °C)

grade

puriss. p.a.

Assay

≥99.0% (T)

form

crystalline

quality

crystallized

impurities

≤0.002% KMnO4-reducing matter (as O)

bp

158 °C (lit.)

mp

~40 °C (lit.)
~40 °C

solubility

water: 850 g/L at 20 °C

density

1.685 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤5 mg/kg
nitrate (NO3-): ≤5 mg/kg
phosphite, hypophosphite (as H3PO3): ≤500 mg/kg
sulfate (SO42-): ≤30 mg/kg

cation traces

As: ≤1 mg/kg
Ca: ≤10 mg/kg
Cd: ≤1 mg/kg
Co: ≤1 mg/kg
Cr: ≤1 mg/kg
Cu: ≤1 mg/kg
Fe: ≤10 mg/kg
K: ≤5 mg/kg
Mg: ≤1 mg/kg
Mn: ≤1 mg/kg
Na: ≤50 mg/kg
Ni: ≤1 mg/kg
Pb: ≤1 mg/kg
Zn: ≤1 mg/kg

SMILES string

OP(O)(O)=O

InChI

1S/H3O4P/c1-5(2,3)4/h(H3,1,2,3,4)

InChI key

NBIIXXVUZAFLBC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The product is crystalline phosphoric acid.

Application

Phosphoric acid has been employed as an oxidation reagent in a study.1 It may be used in the preparation of ammonium phosphate.

Other Notes

Concentrated phosphoric acid is approx. 85% H3PO4 (aqueous).

Caution

product may become solid, becomes liquid on heating to 50°C

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service