Skip to Content
Merck
All Photos(4)

Key Documents

2GFHSNTMGKB

Roche

KAPA2G Fast HotStart PCR Kit

with dNTPs & Mg-free buffers

Synonym(s):

PCR, hotstart PCR

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106300
NACRES:
NA.55

Quality Level

usage

sufficient for 1000 reactions
sufficient for 500 reactions

shelf life

≤18 mo.

feature

Fast PCR
dNTPs included
hotstart

packaging

pkg of 250 U (KK5512)
pkg of 500 U (KK5510)

manufacturer/tradename

Roche

technique(s)

PCR: suitable

input

purified DNA

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

KAPA2G Fast DNA Polymerase is a second-generation (2G) enzyme engineered for higher processivity and speed, offering significantly faster extension rates than wild-type Taq polymerase. In addition to speed, KAPA2G Fast provides higher yields and sensitivity than competitor enzymes across a broad range of targets.
KAPA2G Fast DNA Polymerase Kits are designed for fast PCR, in which total reaction times are 20–70% shorter than those of conventional PCR assays performed with wild-type Taq DNA polymerase. This can be achieved without sacrificing reaction performance, and does not require specialized PCR consumables or thermocyclers.

Application

KAPA2G Fast HotStart® PCR Kit has been used in:
  • Routine PCR
  • Fast PCR
  • Genotyping

Biochem/physiol Actions

DNA fragments generated with KAPA2G Fast DNA Polymerase may be used for routine downstream analyses or applications, including restriction enzyme digestion, cloning and sequencing. Like wild-type Taq, KAPA2G Fast has 5′→3′ polymerase and 5′→3′ exonuclease activities, but no 3′→5′ exonuclease (proofreading) activity. The fidelity of KAPA2G Fast is similar to that of wild-type Taq; it has an error rate of approximately 1 error per 1.7 x 105 nucleotides incorporated.

Features and Benefits

Reduce cycling times up to 75% :
  • Extension times as low as 1 sec per kilobase
  • Broad coverage of both AT- and GC-rich targets

High speed, higher performance :
  • Single copy sensitivity from complex targets
  • Increased yields

Quick Notes:
  • KAPA2G Fast HotStart PCR Kits with dNTPs & Mg-free buffers contain the engineered KAPA2G Fast HotStart DNA Polymerase, developed for fast PCR.
  • Use 1 sec extension time for amplicons <1 kb and 15 sec/kb for longer amplicons, and save 20–70% of total reaction time.
  • No need for specialized instrumentation or PCR consumables.
  • Optimized buffer system offers improved yields,specificity and sensitivity, facilitating efficient primerannealing across a wide range of primer lengths,GC contents, and melting temperatures.
  • Use 0.5 U KAPA2G Fast HotStart DNA Polymerase per 25 μL reaction, or less for smaller volumes.
  • For high reaction efficiency, do not exceed 25 μL reaction volumes.
  • The buffer is available with and without MgCl2 KAPA2G Buffer A contains 1.5 mM MgCl2 at 1X, while KAPA2G Buffer M is Mg-free. Aside from MgCl2 content, these two buffers are identical.
  • Reaction products are 3′-dA-tailed and may be cloned into TA cloning vectors.

Quality

Each batch of KAPA2G Fast DNA Polymerase is confirmed to contain <2% contaminating protein (Agilent Protein 230 Assay). KAPA2G Fast Ready Mix is subjected to stringent quality control tests, is free of contaminating exo- and endonuclease activity, and meets strict requirements with respect to DNA contamination levels.

Preparation Note

Always ensure that the product has been fully thawed and mixed before use. Reagents may be stored at 4°C for short-term use (up to 1 month). Return to -20°C for long-term storage. Provided that all components have been handled carefully and not contaminated, the kit is not expected to be compromised if left (unintentionally) at room temperature for a short period of time (up to 3 days). Long-term storage at room temperature and 4°C is not recommended. Please note that reagents stored at temperatures above -20°C are more prone to degradation when contaminated during use, and therefore storage at such temperatures is at the user′s own risk.

Other Notes

For Research Use Only. Not for use in diagnostic procedures.

Legal Information

HOTSTART is a registered trademark of Molecular BioProducts, Inc.

Kit Components Only

Product No.
Description

  • KAPA2G Fast Standard or HotStart® DNA Polymerase 5 U/μL

  • 5X KAPA2G Buffer M (Mg-free; available with select HotStart kits)

  • MgCl2 25 mM

  • dNTP Mix (optional) 10 mM each dNTP

Pictograms

Exclamation markHealth hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - STOT SE 2

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Investigating the association between polymorphisms in connective tissue growth factor and susceptibility to colon carcinoma.
Ahmad A, et al.
Molecular Medicine Reports, 11(4), 2493-2503 (2015)
Elisa Bergamo et al.
Journal of virological methods, 181(1), 125-130 (2012-01-24)
The potential advantage of using fast PCR to detect human herpesvirus 8 (HHV-8) was tested by running a rapid cycling protocol (5s-steps) in one standard and two fast ramping thermal cyclers to evaluate the performance of 8 different fast reagents.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service