Skip to Content
Merck
All Photos(1)

Key Documents

1.10235

Millipore

SPS agar

GranuCult® plus, suitable for microbiology, for Clostridium spp.

Synonym(s):

Perfringens selective agar, Perfringens selective agar acc. to ANGELOTTI, ANGELOTTI Perfringens selective agar, Sulfite Polymyxin Sulfadiazine Agar

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106213
NACRES:
NA.74

sterility

non-sterile

Quality Level

form

granular

packaging

pkg of 500 g

manufacturer/tradename

GranuCult® plus

storage condition

dry media (Keep tightly closed)

technique(s)

microbiological culture: suitable

color

light brown

pH

6.9-7.1 (35 °C, 40 g/L in H2O, after autoclaving)

solubility

40 g/L

bulk density

580 kg/m3

application(s)

food and beverages
microbiology

storage temp.

15-25°C

suitability

Clostridium perfringens
Clostridium spp.

General description

Sulfite Polymyxin Sulfadiazine (SPS) Agar contains a broad spectrum of nutrients. Most clostridia (including C. perfringens) reduce sulfite to sulfide, which reacts with iron citrate and causes the colonies to turn black. Other sulfite-reducing microorganisms are suppressed by polymyxin and sulfadiazine (sulfapyrimidine). The low sulfite content allows the growth of even sulfite-sensitive clostridia, which exhibit an adequate blackening of the colonies.

Application

SPS agar is suitable for the isolation and enumeration of Clostridium perfringens and Clostridium botulinum in all types of foodstuffs.

Features and Benefits

  • GranuCult® offers superior granulated culture media
  • Safe and sustainable due to reduced risks associated with fine dust and toxic substance inhalation, resulting in a safer work environment
  • Excellent wettability, solubility, and free-flowing properties
  • Convenient, with minimal component separation and clumping, even under warm or humid conditions
  • High batch-to-batch reproducibility
  • Prolonged shelf life of up to five years
  • High number of test strains exceeding all regulatory demands
  • Granulation technology allows many supplements to be included, with no need to add these separately

Analysis Note

Appearance (clearness): clear
Appearance (colour): yellowish-brown
pH-value (25 °C): 6.8 - 7.2
Growth (Clostridium perfringens ATCC 10543): good to very good
Growth (Clostridium perfringens ATCC 13124 (WDCM 00007)): good to very good
Growth (Clostridium sporogenes ATCC 11437): good to very good
Growth (Escherichia coli ATCC 25922 (WDCM 00013)): none to fair
Growth (Pseudomonas aeruginosa ATCC 27853 (WDCM 00025)): none to poor
Blacking (Clostridium perfringens ATCC 10543): +
Blacking (Clostridium perfringens ATCC 13124 (WDCM 00007)): +
Blacking (Clostridium sporogenes ATCC 11437): +
Blacking (Escherichia coli ATCC 25922 (WDCM 00013)): -
Blacking (Pseudomonas aeruginosa ATCC 27853 (WDCM 00025)): -
Incubation: 48 hrs.; 35 °C; aerobic.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

recommended

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service