Key Documents
610037
Avanti
ELISA Frame and Lid (96 well)
Avanti Research - A Croda Brand
About This Item
UNSPSC Code:
41100000
packaging
pkg of 1 ea (610037-1EA)
manufacturer/tradename
Avanti Research - A Croda Brand
storage temp.
room temp
General description
Companion product for Avanti′s ELISA Snoopers.
ELISA Snoopers are 8-well strips pre-coated with a single high-quality lipid species. They can be used to investigate protein-lipid interactions, and characterize and titrate lipid-binding proteins. This format provides the following advantages:
  • quantitative lipid-protein binding studies
  • assemble a 96-well plate using only the lipids of interest for binding studies
  • flexibility - use 8 to 96 wells, and 1 to 12 lipids as needed.
The 8-well strips are available in optically clear, white, or black surfaces for use in colorimetric, luminescent and fluorescent assays, respectively. Avanti Lipid Snoopers ELISA Strips allow multiple sample conditions to be probed simultaneously in a single plate making them a perfect complement to Avanti Lipid Snoopers Nitrocellulose Strips.
Legal Information
Avanti Research is a trademark of Avanti Polar Lipids, LLC
Snoopers is a trademark of Avanti Polar Lipids, LLC