JKA5283
Phosphate-PEG5K
average MN 5,000, NH2, TFA Salt
Synonym(s):
Polyethylene glycol
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
product name
Phosphate-PEG5K-NH2, TFA Salt, average Mn 5000
form
solid
mol wt
average Mn 5000
storage temp.
−20°C
SMILES string
O=P(O)(OCCOCC[NH3+])O.O=CC(F)(F)F
Application
Applications may include: bioconjugation, drug delivery, PEG hydrogel, crosslinker, and surface functionalization
Legal Information
Product of JenKem Technology
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Biomaterials, 168, 1-9 (2018-03-27)
In attempts to explore the role of autophagy in breast cancer metastasis, we here report a tumor-activatable particle (named as "D/PSP@CQ/CaP") with the ability of efficient autophagy inhibition. D/PSP@CQ/CaP was prepared by coprecipitating chloroquine phosphate (CQ) with calcium chloride, in
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service