Skip to Content
Merck
All Photos(2)

Key Documents

GF43125375

Tin

foil, 15mm disks, thickness 0.02mm, 99.75%

Synonym(s):

Tin, SN000180

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Sn
CAS Number:
Molecular Weight:
118.71
MDL number:
UNSPSC Code:
12141745
PubChem Substance ID:
NACRES:
NA.23

Assay

99.75%

form

foil

manufacturer/tradename

Goodfellow 431-253-75

resistivity

11 μΩ-cm, 20°C

diam. × thickness

15 mm × 0.02 mm

bp

2270 °C (lit.)

mp

231.9 °C (lit.)

density

7.310 g/mL at 25 °C (lit.)

SMILES string

[Sn]

InChI

1S/Sn

InChI key

ATJFFYVFTNAWJD-UHFFFAOYSA-N

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Strang et al.
Addiction (Abingdon, England), 92(6), 673-683 (1997-06-01)
The history of heroin smoking and the subsequent development and spread of 'chasing the dragon' are examined. The first heroin smoking originated in Shanghai in the 1920s and involved use of porcelain bowls and bamboo tubes, thereafter spreading across much
Jianan Liu et al.
Animal reproduction science, 134(3-4), 197-202 (2012-08-28)
Cryopreservation of ovarian tissue has been the only effective way of ex situ conservation of female germplasm in avian species. A novel needle-in-straw (NIS) vitrification method was developed to store tissue in straws instead of cryovials. Fragments of ovarian tissue
Andrey A Gurtovenko et al.
The Journal of chemical physics, 130(21), 215107-215107 (2009-06-11)
The electrostatic properties of lipid membranes are of profound importance as they are directly associated with membrane potential and, consequently, with numerous membrane-mediated biological phenomena. Here we address a number of methodological issues related to the computation of the electrostatic
Mercury contamination removed with tin foil.
R Grajower et al.
Operative dentistry, 9(3), 101-104 (1984-01-01)
K A Winship
Adverse drug reactions and acute poisoning reviews, 7(1), 19-38 (1988-01-01)
Inorganic tin salts are poorly absorbed and rapidly excreted in the faeces; as a result they have a low toxicity. Only about 5 per cent is absorbed from the gastrointestinal tract, widely distributed in the body, then excreted by the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service