- Shock and lethality with anthrax edema toxin in rats are associated with reduced arterial responsiveness to phenylephrine and are reversed with adefovir.
Shock and lethality with anthrax edema toxin in rats are associated with reduced arterial responsiveness to phenylephrine and are reversed with adefovir.
American journal of physiology. Heart and circulatory physiology (2017-09-10)
Dante A Suffredini, Yan Li, Wanying Xu, Mahtab Moayeri, Stephen Leppla, Yvonne Fitz, Xizhong Cui, Peter Q Eichacker
PMID28887331
RÉSUMÉ
Although edema toxin (ETx) and lethal toxin (LTx) contribute to
MATÉRIAUX