- Fasciclin-like arabinogalactan proteins, PtFLAs, play important roles in GA-mediated tension wood formation in Populus.
Fasciclin-like arabinogalactan proteins, PtFLAs, play important roles in GA-mediated tension wood formation in Populus.
Scientific reports (2017-07-25)
Haihai Wang, Yanli Jin, Cuiting Wang, Bei Li, Chunmei Jiang, Zhencang Sun, Zhiping Zhang, Fanjing Kong, Hongxia Zhang
PMID28733593
RÉSUMÉ
In Populus, the transcripts of fasciclin-like arabinogalactan proteins (FLAs) are accumulated in tension wood (TW) xylem, however their biological functions in TW formation are largely unknown. In this work, we demonstrated that PtFLA6, one of poplar TW-associated PtFLAs, was abundantly expressed in TW, and mainly localized in differentiating G-fibers. The bended stems of PtFLA6 antisense transgenic poplar showed decreased transcripts of PtFLAs, including PtFLA6, and reduced PtFLA6 like proteins, leading to inhibited TW differentiation and formation. We also showed that gibberellin A3 (GA