Merck
Chemopreventive effects of a low-side-effect antibiotic drug, erythromycin, on mouse intestinal tumors.
Journal of clinical biochemistry and nutrition (2017-06-07)
Takahiro Hamoya, Shingo Miyamoto, Susumu Tomono, Gen Fujii, Ruri Nakanishi, Masami Komiya, Shuya Tamura, Kyoko Fujimoto, Jiro Toshima, Keiji Wakabayashi, Michihiro Mutoh
RÉSUMÉ
It is important to establish effective methods for preventing colorectal cancer because the number of colorectal cancer deaths is increasing. Erythromycin one of the macrolide antibiotics, has been shown to exert pleiotropic effects, such as anti-inflammatory and anti-oxidative effects, on mammalian cells. In the present study, we aimed to evaluate the preventive effects of erythromycin on intestinal carcinogenesis. We first confirmed that erythromycin suppresses the transcriptional activity of nuclear factor-κB and activator protein-1 and the expression of its downstream targets, interleukin-6 and cyclooxygenase-2 in human colon cancer cells. Next, we fed 5-week-old male
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
5HPP-33, ≥98% (HPLC), solid