Merck
Oleanane-type glycosides from Pittosporum tenuifolium "variegatum" and P. tenuifolium "gold star".
Phytochemistry (2017-05-14)
Mayra Rengifo Carrillo, Anne-Claire Mitaine-Offer, Tomofumi Miyamoto, Chiaki Tanaka, Laurent Pouységu, Stéphane Quideau, Luis B Rojas, Carmelo Rosquete Porcar, Marie-Aleth Lacaille-Dubois
RÉSUMÉ
The phytochemical study of two cultivars of Pittosporum tenuifolium Banks & Sol. ex Gaertn, "variegatum" and "gold star", led to the isolation of eight oleanane-type glycosides: seven previously undescribed and a known one. Their aglycons are oxygenated oleanane derivatives as barringtogenol C, camelliagenin A, hederagenin, and 22α-hydroxyoleanolic acid. Their structures were established by 2D NMR spectroscopic techniques and mass spectrometry as 3-O-β-D-galactopyranosyl-(1 → 2)-[α-L-arabinopyranosyl-(1 → 3)]-β-D-glucuronopyranosyl-21-O-angeloyl-22-O-acetylbarringtogenol C, 3-O-β-D-galactopyranosyl-(1 → 2)-[α-L-arabinopyranosyl-(1 → 3)]-β-D-glucuronopyranosyl-21,22-di-O-angeloylbarringtogenol C, 3-O-β-D-galactopyranosyl-(1 → 2)-[α-L-arabinopyranosyl-(1 → 3)]-β-D-glucuronopyranosyl-22-O-angeloylcamelliagenin A, 3-O-β-D-glucopyranosyl-(1 → 2)-[β-D-glucopyranosyl-(1 → 6)]-β-D-glucopyranosyl-22-O-[(6-O-acetyl)-β-D-glucopyranosyl]camelliagenin A, 3-O-β-D-galactopyranosyl-(1 → 2)-[α-L-arabinofuranosyl-(1 → 4)]-β-D-glucuronopyranosylhederagenin 28-O-β-D-glucopyranosyl ester, 3-O-α-L-arabinofuranosyl-(1 → 4)-β-D-glucuronopyranosylhederagenin 28-O-β-D-glucopyranosyl ester, 3-O-β-D-galactopyranosyl-(1 → 2)-[α-L-arabinofuranosyl-(1 → 4)]-β-D-glucuronopyranosyl-22α-hydroxyoleanolic acid 28-O-β-D-glucopyranosyl ester, and the known ilexoside XLIX. These results represent a significative contribution to the chemotaxonomy of the genus Pittosporum, highlighting hederagenin-type saponins as chemotaxonomic markers of P. tenuifolium cultivars.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Thiazolidine, 95%