Merck
Screening of phospholipase A activity and its production by new actinomycete strains cultivated by solid-state fermentation.
PeerJ (2017-07-12)
Priscila Sutto-Ortiz, María de Los Angeles Camacho-Ruiz, Manuel R Kirchmayr, Rosa María Camacho-Ruiz, Juan Carlos Mateos-Díaz, Alexandre Noiriel, Frédéric Carrière, Abdelkarim Abousalham, Jorge A Rodríguez
RÉSUMÉ
Novel microbial phospholipases A (PLAs) can be found in actinomycetes which have been poorly explored as producers of this activity. To investigate microbial PLA production, efficient methods are necessary such as high-throughput screening (HTS) assays for direct search of PLAs in microbial cultures and cultivation conditions to promote this activity. About 200 strains isolated with selected media for actinomycetes and mostly belonging to
MATÉRIAUX
Référence du produit
Marque
Description du produit
2,6-Di-tert-butyl-4-methylphenol, European Pharmacopoeia (EP) Reference Standard