Merck
Acyl Histidines: New N-Acyl Amides from Legionella pneumophila.
Chembiochem : a European journal of chemical biology (2017-01-25)
Thomas Tørring, Stephanie R Shames, Wooyoung Cho, Craig R Roy, Jason M Crawford
RÉSUMÉ
Legionella pneumophila, the causative agent of Legionnaires' disease, is a Gram-negative gammaproteobacterial pathogen that infects and intracellularly replicates in human macrophages and a variety of protozoa. L. pneumophila encodes an orphan biosynthetic gene cluster (BGC) that contains isocyanide-associated biosynthetic genes and is upregulated during infection. Because isocyanide-functionalized metabolites are known to harbor invertebrate innate immunosuppressive activities in bacterial pathogen-insect interactions, we used pathway-targeted molecular networking and tetrazine-based chemoseletive ligation chemistry to characterize the metabolites from the orphan pathway in L. pneumophila. We also assessed their intracellular growth contributions in an amoeba and in murine bone-marrow-derived macrophages. Unexpectedly, two distinct groups of aromatic amino acid-derived metabolites were identified from the pathway, including a known tyrosine-derived isocyanide and a family of new N-acyl-l-histidine metabolites.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
cis-2-Decenoic acid, ≥95.0% (HPLC)