Merck
Immunoassays of chemically modified polysaccharides, glycans in glycoproteins and ribose in nucleic acids.
Analytica chimica acta (2017-01-16)
Slađana Strmečki, Mojmír Trefulka, Pavlína Zatloukalová, Michal Durech, Borivoj Vojtesek, Emil Paleček
RÉSUMÉ
Glycosylation of proteins plays an important role in health and diseases. At present new simple and inexpensive methods of glycoprotein analysis are sought. We developed a monoclonal antibody Manost 2.1 in mice after immunization with the adduct of mannan with Os(VI)temed complex (temed is N,N,N',N'-tetramethylethylenediamine). The specificity of this antibody to different biomolecules treated with Os(VI)temed was tested using dot blot immunoassay. Manost 2.1 showed specificity toward Os(VI)temed-modified polysaccharides, glycoproteins and ribonucleotide at the 3'-end in DNA. The antibody recognized neither the unmodified compounds nor the non-glycosylated proteins treated with Os(VI)temed. We also performed western blotting and Coomassie silver blue staining of mixtures of biomacromolecules treated with Os(VI)temed and identified specifically the modified glycoproteins. The immunochemical method using Manost 2.1 was compared with electrochemical analyses based on redox signals of the Os(VI)temed adducts, with similar results in terms of sensitivity. This new antibody-based approach opens the door for rapid and inexpensive analysis of glycans and glycoproteins in various scientific and medical fields, including cancer research and the future application of glycoprotein detection in clinical practice.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
1,10-Phenanthroline monohydrate, ACS reagent, 99%