Merck
Direct C-H arylation of (hetero)arenes with aryl iodides via rhodium catalysis.
Journal of the American Chemical Society (2006-09-07)
Shuichi Yanagisawa, Tomoko Sudo, Ryoji Noyori, Kenichiro Itami
RÉSUMÉ
A new method for the catalytic C-H arylation of heteroarenes and arenes that manifests high activity paired with reasonably broad scope was developed. Under the catalytic influence of RhCl(CO){P[OCH(CF3)2]3}2 and Ag2CO3, the direct C-H arylation of heteroarenes/arenes with aryl/heteroaryl iodides took place to afford a range of biaryls in good to excellent yields with high regioselectivity. Thiophenes, furans, pyrroles, indoles, and alkoxybenzenes are applicable in this arylation protocol.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
2,2′-Bithiophene, 99%