Merck
Effect of tumor-associated macrophages on gastric cancer stem cell in omental milky spots and lymph node micrometastasis.
International journal of clinical and experimental pathology (2016-01-30)
Chi Zhang, Xiang Hu, Xiao-Yu Liu, Pin Liang, Jian Zhang, Liang Cao, Zheng-Lin Wang, Huan-Ran Liu, Xun-Guo Yin, Cheng-Yong Dong, Li-Ming Wang
RÉSUMÉ
We observed whether the effect of tumor-associated macrophages on gastric cancer stem cell in omental milky spots and lymph nodes micrometastasis and research its possible mechanism. Macrophage THP-1 cells and Human gastric adenocarcinoma SGC-7901 cells were collectively cultivated in vivo. We found macrophage could suppress the proliferation and accelerated cell death of MFC cell. Meanwhile, these effects may be concerned with many signaling pathways, and we detected MCP-1 and COX-2 miRNA expressions, PGE-2 release levels, IL-4 and IL-10 activities, and TGF-β, IFN-γ, VEGF, MMP-2 and MMP-9 protein expressions in collectively cultivated cell. We found that MCP-1 and COX-2 miRNA expressions, and PGE-2 release levels were suppressed, IL-4 activity was inhibited and IL-10 activity was activated in collectively cultivated cell. Meanwhile, TGF-β, MMP-2 and MMP-9 protein expressions were inhibited and IFN-γ and VEGF protein expressions were activated in collectively cultivated cell. Taken together, these results suggest that the effect of tumor-associated macrophages on gastric cancer stem cell in omental milky spots and lymph nodes micrometastasis via COX-2/PGE-2/TGF-β/VEGF signal pathways.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
2-Methyl-1,3-thiazole, AldrichCPR