Merck
A network of heterochronic genes including Imp1 regulates temporal changes in stem cell properties.
eLife (2013-11-07)
Jinsuke Nishino, Sunjung Kim, Yuan Zhu, Hao Zhu, Sean J Morrison
RÉSUMÉ
Stem cell properties change over time to match the changing growth and regeneration demands of tissues. We showed previously that adult forebrain stem cell function declines during aging because of increased expression of let-7 microRNAs, evolutionarily conserved heterochronic genes that reduce HMGA2 expression. Here we asked whether let-7 targets also regulate changes between fetal and adult stem cells. We found a second let-7 target, the RNA binding protein IMP1, that is expressed by fetal, but not adult, neural stem cells. IMP1 expression was promoted by Wnt signaling and Lin28a expression and opposed by let-7 microRNAs. Imp1-deficient neural stem cells were prematurely depleted in the dorsal telencephalon due to accelerated differentiation, impairing pallial expansion. IMP1 post-transcriptionally inhibited the expression of differentiation-associated genes while promoting the expression of self-renewal genes, including Hmga2. A network of heterochronic gene products including Lin28a, let-7, IMP1, and HMGA2 thus regulates temporal changes in stem cell properties. DOI: http://dx.doi.org/10.7554/eLife.00924.001.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Anticorps anti-nestine, clone rat-401, clone rat-401, Chemicon®, from mouse
Sigma-Aldrich
Anticorps anti-TCF-4, clone 6H5-3, clone 6H5-3, Upstate®, from mouse