Merck
The concept of superactive esters. Could peptide synthesis be improved by inventing superactive esters?
International journal of peptide and protein research (1994-03-01)
Z J Kamiński
RÉSUMÉ
According to the concept presented, esters forming an amide (peptide) bond by the mechanism SN#DN or SN#*DN involving fast decay of the tetrahedral intermediate may behave as 'superactive acylating reagents'. These should render coupling involving less reactive substrates, i.e. sterically hindered or 'difficult coupling sequences', much faster and more uniform than classic active esters. In extremely cases this advantage could be significant, and the calculated increase in time required for 99.9% coupling substrates 6 pKa units less reactive than the standard ones reaches 2,512,000 tau 1/2 units for classic active esters, but only 631 tau 1/2 units for reaction involving 'superactive esters'. The postulated change of mechanism is expected for esters bearing a leaving group which is able to undergo an additional, synchronous, energetically favored process accompanying its departure, as has been observed in the case of triazine esters. Some advantages of triazine superactive esters in the condensation of sterically hindered substrates are demonstrated.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Trimethylacetic anhydride, 99%