Merck
Upregulation of IGF-1R expression during neoadjuvant therapy predicts poor outcome in breast cancer patients.
PloS one (2015-02-14)
Sandra Heskamp, Otto C Boerman, Janneke D M Molkenboer-Kuenen, Carla A Wauters, Luc J A Strobbe, Caroline M P W Mandigers, Peter Bult, Wim J G Oyen, Winette T A van der Graaf, Hanneke W M van Laarhoven
RÉSUMÉ
The insulin-like growth factor 1 receptor (IGF-1R) may be involved in the development of resistance against conventional cancer treatment. The aim of this study was to assess whether IGF-1R expression of breast tumors changes during neoadjuvant therapy and to study whether these changes were associated with survival. Paraffin embedded tumor tissue was collected from pretreatment biopsies and surgical resections of 62 breast cancer patients who were treated with neoadjuvant chemotherapy or endocrine therapy. IGF-1R expression was determined immunohistochemically and compared before and after treatment. High membranous IGF-1R expression at diagnosis correlated significantly with ER positivity, low tumor stage (stage I/II) and longer overall survival (p < 0.05). After neoadjuvant treatment, membranous IGF-1R expression remained the same in 41 (65%) tumors, was upregulated in 11 (18%) tumors and downregulated in 11 (18%) tumors. Changes in membranous IGF-1R expression were associated with overall survival (log-rank test: p = 0.013, multivariate cox-regression: p = 0.086). Mean overall survival time for upregulation, no change, and downregulation in IGF-1R expression was 3.0 ± 0.5 years, 7.3 ± 1.0 years and 15.0 ± 1.8 years, respectively. Changes in other parameters were not significantly associated with survival. Neoadjuvant therapy can induce changes in IGF-1R expression. Upregulation of IGF-1R expression after neoadjuvant treatment is a poor prognostic factor in breast cancer patients, providing a rationale for incorporating anti-IGF-1R drugs in the management of these patients.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Insulin-like Growth Factor-I Receptor human, ≥95% (SDS-PAGE), recombinant, expressed in NSO cells, lyophilized powder