Merck
Structural identification and characterization of potential impurities of pantoprazole sodium.
Journal of pharmaceutical and biomedical analysis (2007-07-17)
Ganta Madhusudhan Reddy, B Vijaya Bhaskar, P Pratap Reddy, S Ashok, P Sudhakar, J Moses Babu, K Vyas, K Mukkanti
RÉSUMÉ
Six impurities in pantoprazole sodium bulk drug substance were detected by a simple high performance liquid chromatographic method (HPLC) whose area percentage ranged from approximately 0.05 to 0.34%. Liquid chromatography-mass spectrometry (LC-MS) was performed to identify the molecular weight of the impurities. A thorough study was undertaken to characterize these impurities. These impurities were synthesized, subsequently characterized and were co-injected with the sample containing impurities and found the retention time match of the spiked impurities. Based on their spectral data (IR, NMR and MS), these impurities were characterized as; 5-(difluoromethoxy)-2-[[(3,4-dimethoxy-2-pyridinyl)methyl]thio]-1H-benzimidazole (Impurity-I); 5-(difluoromethoxy)-2-[[(3,4-dimethoxy-2-pyridinyl)methyl]sulfonyl]-1H-benzimidazole (Impurity-II); 5-(difluoromethoxy)-2-[[(3,4-dimethoxy-1-oxide-2-pyridinyl)methyl]sulfonyl]-1H-benzimidazole (Impurity-III); 5-(difluoromethoxy)-2-[[(3,4-dimethoxy-2-pyridinyl)methyl]thio]-1-((3,4-dimethoxy-2-pyridinyl)methyl)-1H-benzimidazole (Impurity-IV); 5-(difluoromethoxy)-2-[[(3,4-dimethoxy-2-pyridinyl)methyl]sulfinyl]-1-((3,4-dimethoxy-2-pyridinyl)methyl)-1H-benzimidazole (Impurity-V); 5-(difluoromethoxy)-2-[[(3,4-dimethoxy-1-oxide-2-pyridinyl)methyl]sulfinyl]-1H-benzimidazole (Impurity-VI). The formation of these impurities was proposed. The structure of the Impurity-II was unambiguously confirmed by single crystal X-ray diffraction (XRD) studies.
MATÉRIAUX
Référence du produit
Marque
Description du produit
USP
Pantoprazole Related Compound C, United States Pharmacopeia (USP) Reference Standard