Merck
GM130 and GRASP65-dependent lateral cisternal fusion allows uniform Golgi-enzyme distribution.
Nature cell biology (2006-02-21)
Manojkumar A Puthenveedu, Collin Bachert, Sapna Puri, Frederick Lanni, Adam D Linstedt
RÉSUMÉ
The mammalian Golgi apparatus exists as stacks of cisternae that are laterally linked to form a continuous membrane ribbon, but neither the molecular requirements for, nor the purpose of, Golgi ribbon formation are known. Here, we demonstrate that ribbon formation is mediated by specific membrane-fusion events that occur during Golgi assembly, and require the Golgi proteins GM130 and GRASP65. Furthermore, these GM130 and GRASP65-dependent lateral cisternal-fusion reactions are necessary to achieve uniform distribution of enzymes in the Golgi ribbon. The membrane continuity created by ribbon formation facilitates optimal processing conditions in the biosynthetic pathway.