Merck
  • Clinical expression and new SPINK5 splicing defects in Netherton syndrome: unmasking a frequent founder synonymous mutation and unconventional intronic mutations.
Clinical expression and new SPINK5 splicing defects in Netherton syndrome: unmasking a frequent founder synonymous mutation and unconventional intronic mutations.
The Journal of investigative dermatology (2011-11-18)
Matthieu Lacroix, Laetitia Lacaze-Buzy, Laetitia Furio, Elodie Tron, Manthoula Valari, Gerda Van der Wier, Christine Bodemer, Anette Bygum, Anne-Claire Bursztejn, George Gaitanis, Mauro Paradisi, Alexander Stratigos, Lisa Weibel, Céline Deraison, Alain Hovnanian
RÉSUMÉ
Netherton syndrome (NS) is a severe skin disease caused by loss-of-function mutations in SPINK5 (serine protease inhibitor Kazal-type 5) encoding the serine protease inhibitor LEKTI (lympho-epithelial Kazal type-related inhibitor). Here, we disclose new SPINK5 defects in 12 patients, who presented a clinical triad suggestive of NS with variations in inter- and intra-familial disease expression. We identified a new and frequent synonymous mutation c.891C>T (p.Cys297Cys) in exon 11 of the 12 NS patients. This mutation disrupts an exonic splicing enhancer sequence and causes out-of-frame skipping of exon 11. Haplotype analysis indicates that this mutation is a founder mutation in Greece. Two other new deep intronic mutations, c.283-12T>A in intron 4 and c.1820+53G>A in intron 19, induced partial intronic sequence retention. A new nonsense c.2557C>T (p.Arg853X) mutation was also identified. All mutations led to a premature termination codon resulting in no detectable LEKTI on skin sections. Two patients with deep intronic mutations showed residual LEKTI fragments in cultured keratinocytes. These fragments retained some functional activity, and could therefore, together with other determinants, contribute to modulate the disease phenotype. This new founder mutation, the most frequent mutation described in European populations so far, and these unusual intronic mutations, widen the clinical and molecular spectrum of NS and offer new diagnostic perspectives for NS patients.