Merck
Stereoselective α-quaternization of 3-methoxycycloalk-2-enones via 1,4-diastereoinduction of alkoxy dienolates.
The Journal of organic chemistry (2012-01-03)
Gamal A I Moustafa, Yasumasa Kamada, Tetsuaki Tanaka, Takehiko Yoshimitsu
RÉSUMÉ
The alkylation of dienolates generated from 3-methoxycycloalk-2-enones having a 3'-hydroxyl alkenyl chain provides the corresponding quaternized cycloalkenones in a highly diastereoselective manner. The high degree of stereocontrol in the α-quaternization possibly implies intervention of a rigid chelating transition state that allows an efficient 1,4-asymmetric induction to take place.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
3-Methoxy-2-cyclopenten-1-one, 99%