Merck
Synthesis of quinolines from 3-formylchromone.
The Journal of organic chemistry (2008-07-03)
Andrey S Plaskon, Sergey V Ryabukhin, Dmitriy M Volochnyuk, Konstantin S Gavrilenko, Alexander N Shivanyuk, Andrey A Tolmachev
RÉSUMÉ
A facile and versatile procedure for the synthesis of 3-(2-hydroxybenzoyl)quinolines and 7H-chromeno[3,2-c]quinolin-7-ones was elaborated on the basis of TMSCl-mediated recyclization of 3-formylchromone with various anilines. Limitations and scope of this methodology were established, and a possible mechanism for the heterocyclizations was proposed.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
3-Formylchromone, 97%