Merck
The first oxazoline adduct of Zn(acac)(2): bis-(acetyl-acetonato-κO,O')(2-phenyl-2-oxazoline-κN)zinc(II).
Acta crystallographica. Section E, Structure reports online (2008-01-01)
Ignacio Del Río, Robert A Gossage
RÉSUMÉ
The title material, [Zn(C(5)H(7)O(2))(2)(C(9)H(9)NO)], was synthesized by the treatment of bis-(acetyl-acetonato)zinc(II) monohydrate with 2-phenyl-2-oxazoline. The Zn atom is coordinated by two chelating acetyl-acetonate groups and one oxazoline ligand in the apical position of a slightly distorted square-pyramidal metal-ligand geometry.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
2-Phenyl-2-oxazoline, 99%