Merck
  • Proteomic-based identification of the APC-binding protein EB1 as a candidate of novel tissue biomarker and therapeutic target for colorectal cancer.
Proteomic-based identification of the APC-binding protein EB1 as a candidate of novel tissue biomarker and therapeutic target for colorectal cancer.
Journal of proteomics (2012-06-28)
Yutaka Sugihara, Hirokazu Taniguchi, Ryoji Kushima, Hitoshi Tsuda, Daisuke Kubota, Hiroshi Ichikawa, Kano Sakamoto, Yukiko Nakamura, Takeshi Tomonaga, Shin Fujita, Tadashi Kondo
RÉSUMÉ
Novel candidates of biomarker and therapeutic target in colorectal cancer (CRC) were investigated using a proteomic approach. The proteome of normal colorectal epithelial tissues was compared with that of the tumor ones in 59 CRC patients using two-dimensional difference gel electrophoresis. Of 3458 protein spots, 110 exhibited statistically significant (p<0.01) differences in intensity (more than 2.5-folds) between the normal and tumor tissue groups. Of 67 unique gene products that were identified for 105 of the 110 protein spots, we focused on the higher expression of the adenoma polyposis coli-binding protein EB1 (EB1). EB1 was originally discovered as a binding protein of APC, which is a tumor suppressor gene product, and the expression of EB1 has been associated with poor prognosis in several malignancies but not in CRC. Immunohistochemical analysis of the 132 CRC cases revealed that EB1 was overexpressed in tumor cells in correlation with poor prognosis. Suppression of EB1 by RNAi inhibited CRC cell proliferation and invasion. In this study, the overexpression of EB1 in CRC tissues correlating with prognosis, and its functional contribution to the malignant phenotypes of CRC cells are described. The present findings indicate that EB1 is a potential biomarker and therapeutic target in CRC.