Merck
A general approach to mechanism in multiproduct reactions: product-specific intermolecular kinetic isotope effects.
Journal of the American Chemical Society (2014-04-12)
Shuhuai Xiang, Matthew P Meyer
RÉSUMÉ
Here we report a general method for the measurement of (13)C kinetic isotope effects at natural abundance for reactions that yield two or more products concurrently. We use, as an example, a recently reported Co-catalyzed reaction between cyclopentene and 1-phenyl-1-propyne. High-precision intermolecular (13)C isotope effects are reported for both the formal [2+2] cycloaddition (major) and Alder-ene (minor) reaction products. Mechanistic possibilities that are in accord with observed isotope effect measurements are discussed.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
1-Phenyl-1-propyne, 99%