Merck
  • Combined speciation analysis by X-ray absorption near-edge structure spectroscopy, ion chromatography, and solid-phase microextraction gas chromatography-mass spectrometry to evaluate biotreatment of concentrated selenium wastewaters.
Combined speciation analysis by X-ray absorption near-edge structure spectroscopy, ion chromatography, and solid-phase microextraction gas chromatography-mass spectrometry to evaluate biotreatment of concentrated selenium wastewaters.
Environmental science & technology (2010-12-25)
Markus Lenz, Eric D van Hullebusch, François Farges, Sergei Nikitenko, Philippe F X Corvini, Piet N L Lens
RÉSUMÉ
In this study we evaluate the potential of anaerobic granular sludge as an inoculum for the bioremediation of selenium-contaminated waters using species-specific analytical methods. Solid species formed by microbial reduction were investigated using X-ray absorption near-edge structure (XANES) spectroscopy at the selenium K-edge. Furthermore, dissolved selenium species were specifically determined by ion chromatography (IC) and solid-phase microextraction gas chromatography-mass spectrometry (SPME-GC-MS). Least-squares linear combination of the XANES spectra for samples incubated with the highest selenate/selenite concentrations (10(-3) M) show the predominance of elemental selenium and a Se(-I) selenide, such as ferroselite, the thermodynamically most stable iron selenide. In contrast, elemental selenium and Se(-II) selenides are the main species detected at the lower selenate/selenite concentrations. In each repeated fed batch incubation, most aqueous selenite anions were converted into solid selenium species, regardless of the type of electron donor used (acetate or H(2)/CO(2)) and the selenium concentration applied. On the other hand, at higher concentrations of selenate (10(-4) and 10(-3) M), significant amounts of the oxyanion remained unconverted after consecutive incubations. SPME-GC-MS demonstrated selenium alkylation with both electron donors investigated, as dimethyl selenide (DMSe) and dimethyl diselenide (DMDSe). Selenite was even more alkylated in the presence of H(2)/CO(2) (maximum 2156 μg of Se/L of DMSe + DMDSe) as compared to acetate (maximum 50 μg of Se/L). In contrast, selenate was less alkylated using both electron donors (maximum 166 and 3 μg of Se/L, respectively). The high alkylation potential for selenite limits its bioremediation in selenium laden waters involving H(2)/CO(2) as the electron donor despite the fact that nontoxic elemental selenium and thermodynamically stable metal selenide species are formed.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Dimethyl diselenide, 96%