Merck
  • Conversion of phenol derivatives to hydroxylated products by phenol hydroxylase from Trichosporon cutaneum. A comparison of regioselectivity and rate of conversion with calculated molecular orbital substrate characteristics.
Conversion of phenol derivatives to hydroxylated products by phenol hydroxylase from Trichosporon cutaneum. A comparison of regioselectivity and rate of conversion with calculated molecular orbital substrate characteristics.
European journal of biochemistry (1995-01-15)
S Peelen, I M Rietjens, M G Boersma, J Vervoort
RÉSUMÉ
This study describes the regioselective hydroxylation and the rates of conversion of a series of fluorinated phenol derivatives by phenol hydroxylase from the yeast Trichosporon cutaneum. The natural logarithm of the kcat value for the conversion of the phenolic substrates correlates with the calculated energy of the reactive electrons in the highest occupied molecular orbital of the substrate (r = 0.85). This observation supports the hypothesis that at physiological pH (7.6) and 25 degrees C, in the absence of monovalent anions, the nucleophilic attack of the electrons in the highest occupied molecular orbital of the substrate on the C(4a)-hydroperoxyflavin enzyme intermediate is of major importance in determining the overall rate of catalysis. Results from 19F-NMR analysis of the incubation mixtures demonstrate for phenols with two identical ortho substituents, that the ortho position which becomes preferentially hydroxylated is the one with the highest density of the reactive electrons in the highest occupied molecular orbital. A halogen substituent at a meta position decreases the chances for hydroxylation at the adjacent ortho position further than expected on the basis of the calculated reactivity. This result indicates a contribution of a protein/substrate dipolar interaction, influencing the time-averaged orientation of the substrate with respect to the reactive C(4a)-hydroperoxyflavin intermediate.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
4-Chloro-3-fluorophenol, 98%