Merck
Differentially expressed proteins in malignant and benign adrenocortical tumors.
PloS one (2014-02-06)
Hanna Kjellin, Henrik Johansson, Anders Höög, Janne Lehtiö, Per-Johan Jakobsson, Magnus Kjellman
RÉSUMÉ
We have compared the microsomal protein composition of eight malignant and six benign adrenocortical tumors with proteomic methods. IGF2 had increased level in the malignant tumors, confirming previous microarray studies on the same material. Aldolase A, a glycolytic enzyme, also showed increased levels in the malignant tissue compared to the benign. Additionally, several proteins belonging to complex I in the mitochondrial respiration chain showed decreased levels in the malignant tissue. Taken together, this may indicate a shift in energy metabolism where glycolysis may be favored over tight coupling of glycolysis and mitochondrial respiration, a phenomenon known as the Warburg effect. One of the complex I proteins that showed decreased levels in the malignant tissue was GRIM-19. This protein has been suggested as a tumor suppressive protein by being a negative regulator of STAT3. In summary, an analysis of the microsomal proteome in adrenocortical tumors identifies groups of proteins as well as specific proteins differentially expressed in the benign and malignant forms. These proteins shed light on the biology behind malignancy and could delineate future drug targets.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Anti-SHMT2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab2