Merck
Postablation neutrophil/lymphocyte ratio correlates with arrhythmia recurrence after catheter ablation of lone atrial fibrillation.
Chinese medical journal (2014-03-14)
Xueyuan Guo, Sen Zhang, Xianliang Yan, Yingwei Chen, Ronghui Yu, Deyong Long, Caihua Sang, Xin Du, Jianzeng Dong, Changsheng Ma
RÉSUMÉ
Neutrophil/lymphocyte ratio (NLR) has been proposed as a novel marker of systemic inflammation and oxidative stress. The objective of this study was to ascertain the relationship between levels of NLR and recurrence of lone atrial fibrillation (AF) after catheter ablation. A total of 379 lone AF patients who underwent catheter ablation were enrolled in the study. The NLR before and after catheter ablation was determined. Cox regression analyses were used to estimate the relationship between NLR and the recurrence of lone AF. After a mean follow-up of (30.5 ± 5.3) months, 124 (32.7%) patients had AF recurrences. The patients who developed AF recurrence had a higher postablation NLR (post-NLR) than patients with no recurrence (5.74 ± 1.55 vs. 4.66 ± 1.27, P < 0.001). Multivariate Cox regression analysis revealed that post-NLR (hazard ratio (HR) 1.514, 95% confidence interval (CI) 1.364-1.680, P < 0.001), left atrium diameter (HR 1.035, 95% CI 1.001-1.071, P = 0.04) and body mass index (HR 1.028, 95% CI 1.002-1.054, P = 0.03) were independent predictors of AF recurrence. Using a cut-off level of 5.15, post-NLR predicted AF recurrence with a sensitivity of 73% and specificity of 67%. Our results indicate that an elevated post-NLR is associated with a high rate of lone AF recurrence. A simple measurement of NLR may help us to identify high-risk patients who need pharmacologic intervention to prevent recurrence.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
CKI-7 dihydrochloride, ≥98% (HPLC), solid