Merck
Ionized o-, m-, and p-difluorobenzene dissociate through ring-opened intermediates: a TPEPICO investigation.
Journal of the American Society for Mass Spectrometry (2008-10-18)
Anne-Marie Boulanger, David M P Holland, David A Shaw, Paul M Mayer
RÉSUMÉ
Threshold photoelectron photoion coincidence (TPEPICO) experiments have shown that o-, m-, and p-difluorobenzene ions dissociate via a common, ring-opened intermediate and not via ionized p-difluorobenzene. Rice-Ramsperger-Kassel-Marcus (RRKM) modeling of the experimental breakdown curves yields activation energies for the initial isomerization of 4.48 +/- 0.05, 4.55 +/- 0.05, and 4.68 +/- 0.05 eV for o-, m-, and p-difluorobenzene, respectively. These values place each ion at a similar absolute energy and thus similar transition states. A large positive DeltaS(double dagger) for each ion (ca 100 J K(-1) mol(-1)) suggests a ring-opened structure for these transition states.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
1,2-Difluorobenzene, 98%