- Study of local anaesthetics. Part 133. Determination of diperodon in blood serum in vitro by the HPLC method.
Study of local anaesthetics. Part 133. Determination of diperodon in blood serum in vitro by the HPLC method.
Acta poloniae pharmaceutica (1996-05-01)
J Cizmárik, J Lehotay, K Hromuláková
PMID8960289
RÉSUMÉ
A local anaesthetics diperodon by the action of hydrolazes in blood serum is decomposed. A method for its determination in blood serum in vitro has been worked out. It was found that during the first 2 hours it decomposes rather rapidly. During next 6 hours the rate of decomposition is decreased, and after 8 hours the blood serum contained yet about 33% of not decomposed diperodon. The separation of diperodon has been carried out in the chromatographic system which consists of 70% methanol in mobile phase in water with sodium acetate (6.8 g/l, pH = 7.4) and chemically bounded phase C18. The capacity ratio of diperodon has been 8.1, the limit of determination is 5 micrograms/ml, and the exctraction recovery is 70%.