Merck
Bisantrene in relapsed and refractory acute myelogenous leukemia.
Leukemia & lymphoma (1993-02-01)
A Spadea, M C Petti, M A Aloespiriti, G Avvisati, C De Gregoris, P Fazi, R Latagliata, S Amadori, F Mandelli
RÉSUMÉ
Because of the lack of standard treatment in refractory and relapsed acute myelogenous leukemia (AML) several new drugs have been employed alone to evaluate their efficacy in this peculiar category of patient. Bisantrene, a new anthracene bishydrazone derivative, has shown antileukemic effect in phase I and II clinical trials with acceptable extrahaematological toxicity. Seven patients (six males and one female, median age 41.8 years) received Bisantrene (250 mg/sqm/daily 1-7) as a single agent in relapsed or refractory leukemia. 5 out of 7 patients achieved complete remission, one attained partial remission and one was resistant. However, haematological toxicity was severe with prolonged myelosuppression. Hepatic toxicity was the main extrahaematological side effect and occurred in 3 of 7 patients, however all of them recovered within 40 days. No cardiovascular dysfunction occurred although all the patients had been heavily previously treated with anthracyclines. Our data confirm that Bisantrere is active in relapsed and refractory AML and suggest the need for larger clinical trials to better evaluate its efficacy.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Bisantrene dihydrochloride, ≥98% (HPLC)