- Determination of trimethoprim, sulphamethoxazole and its N4-acetyl metabolite in biological fluids by high-performance liquid chromatography.
Determination of trimethoprim, sulphamethoxazole and its N4-acetyl metabolite in biological fluids by high-performance liquid chromatography.
Journal of chromatography (1983-05-13)
O Spreux-Varoquaux, J P Chapalain, P Cordonnier, C Advenier, M Pays, L Lamine
PMID6603465
RÉSUMÉ
A normal-phase high-performance liquid chromatographic method was developed to determine therapeutic concentrations of trimethoprim, sulphamethoxazole, and its N4-acetyl derivative in biological fluids. The compounds are extracted at pH 6.2 using ethyl acetate--chloroform in a single extraction. The detection limit is 15 ng/ml for trimethoprim, 20 ng/ml for sulphamethoxazole, and 10 ng/ml for its N4-acetyl metabolite. The method is rapid, sensitive, precise, and well suited to clinical pharmacokinetic investigations.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sulfamethoxazole impurity A, European Pharmacopoeia (EP) Reference Standard