Merck
Deoxyribonuclease I is essential for DNA fragmentation induced by gamma radiation in mice.
Radiation research (2009-09-24)
Eugene O Apostolov, Izoumroud Soultanova, Alena Savenka, Osman O Bagandov, Xiaoyan Yin, Anna G Stewart, Richard B Walker, Alexei G Basnakian
RÉSUMÉ
Gamma radiation is known to induce cell death in several organs. This damage is associated with endonuclease-mediated DNA fragmentation; however, the enzyme that produces the latter and is likely to cause cell death is unknown. To determine whether the most abundant cytotoxic endonuclease DNase I mediates gamma-radiation-induced tissue injury, we used DNase I knockout mice and zinc chelate of 3,5-diisopropylsalicylic acid (Zn-DIPS), which, as we show, has DNase I inhibiting activity in vitro. The study demonstrated for the first time that inactivation or inhibition of DNase I ameliorates radiation injury to the white pulp of spleen, intestine villi and bone marrow as measured using a quantitative TUNEL assay. The spleen and intestine of DNase I knockout mice were additionally protected from radiation by Zn-DIPS, perhaps due to the broad radioprotective effect of the zinc ions. Surprisingly, the main DNase I-producing tissues such as the salivary glands, pancreas and kidney showed no effect of DNase I inactivation. Another unexpected observation was that even without irradiation, DNA fragmentation and cell death were significantly lower in the intestine of DNase I knockout mice than in wild-type mice. This points to the physiological role of DNase I in normal cell death in the intestinal epithelium. In conclusion, our results suggested that DNase I-mediated mechanism of DNA damage and subsequent tissue injury are essential in gamma-radiation-induced cell death in radiosensitive organs.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
3,5-Diisopropyl-2-hydroxybenzoic acid, 98%