Merck
  • 5-hydroxyconiferyl aldehyde modulates enzymatic methylation for syringyl monolignol formation, a new view of monolignol biosynthesis in angiosperms.
5-hydroxyconiferyl aldehyde modulates enzymatic methylation for syringyl monolignol formation, a new view of monolignol biosynthesis in angiosperms.
The Journal of biological chemistry (2000-02-29)
L Li, J L Popko, T Umezawa, V L Chiang
RÉSUMÉ
S-Adenosyl-L-methionine-dependent caffeate O-methyltransferase (COMT, EC 2.1.1.6) has traditionally been thought to catalyze the methylation of caffeate and 5- hydroxyferulate for the biosynthesis of syringyl monolignol, a lignin constituent of angiosperm wood that enables efficient lignin degradation for cellulose production. However, recent recognition that coniferyl aldehyde prevents 5-hydroxyferulate biosynthesis in lignifying tissue, and that the hydroxylated form of coniferyl aldehyde, 5-hydroxyconiferyl aldehyde, is an alternative COMT substrate, demands a re-evaluation of the role of COMT during monolignol biosynthesis. Based on recombinant aspen (Populus tremuloides) COMT enzyme kinetics coupled with mass spectrometry analysis, this study establishes for the first time that COMT is in fact a 5-hydroxyconiferyl aldehyde O-methyltransferase (AldOMT), and that 5-hydroxyconiferyl aldehyde is both the preferred AldOMT substrate and an inhibitor of caffeate and 5-hydroxyferulate methylation, as measured by K(m) and K(i) values. 5-Hydroxyconiferyl aldehyde also inhibited the caffeate and 5-hydroxyferulate methylation activities of xylem proteins from various angiosperm tree species. The evidence that syringyl monolignol biosynthesis is independent of caffeate and 5-hydroxyferulate methylation supports our previous discovery that coniferyl aldehyde prevents ferulate 5-hydroxylation and at the same time ensures a coniferyl aldehyde 5-hydroxylase (CAld5H)-mediated biosynthesis of 5-hydroxyconiferyl aldehyde. Together, our results provide conclusive evidence for the presence of a CAld5H/AldOMT-catalyzed coniferyl aldehyde 5-hydroxylation/methylation pathway that directs syringyl monolignol biosynthesis in angiosperms.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
3,4-Dihydroxy-5-methoxycinnamic acid, ≥95.0% (HPLC)