Merck
  • A branched electrode based electrochemical platform: towards new label-free and reagentless simultaneous detection of two biomarkers.
A branched electrode based electrochemical platform: towards new label-free and reagentless simultaneous detection of two biomarkers.
Chemical communications (Cambridge, England) (2012-11-22)
Fen-Ying Kong, Bi-Yi Xu, Ying Du, Jing-Juan Xu, Hong-Yuan Chen
RÉSUMÉ
A branched electrode platform was proposed for label-free and reagentless simultaneous tumor markers detection based on different redox substrates.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Iron(III) ferrocyanide
Sigma-Aldrich
Tin(IV) oxide, nanopowder, ≤100 nm avg. part. size
Sigma-Aldrich
Tin(IV) oxide, −325 mesh, 99.9% trace metals basis
Sigma-Aldrich
Tin(IV) oxide, ≥99.99% trace metals basis