Merck
Gustatory receptors required for avoiding the insecticide L-canavanine.
The Journal of neuroscience : the official journal of the Society for Neuroscience (2012-01-27)
Youngseok Lee, Min Jung Kang, Jaewon Shim, Chae Uk Cheong, Seok Jun Moon, Craig Montell
RÉSUMÉ
Insect survival depends on contact chemosensation to sense and avoid consuming plant-derived insecticides, such as L-canavanine. Members of a family of ∼60 gustatory receptors (GRs) comprise the main peripheral receptors responsible for taste sensation in Drosophila. However, the roles of most Drosophila GRs are unknown. In addition to GRs, a G protein-coupled receptor, DmXR, has been reported to be required for detecting L-canavanine. Here, we showed that GRs are essential for responding to L-canavanine and that flies missing DmXR displayed normal L-canavanine avoidance and L-canavanine-evoked action potentials. Mutations disrupting either Gr8a or Gr66a resulted in an inability to detect L-canavanine. We found that L-canavanine stimulated action potentials in S-type sensilla, which were where Gr8a and Gr66a were both expressed, but not in Gr66a-expressing sensilla that did not express Gr8a. L-canavanine-induced action potentials were also abolished in the Gr8a and Gr66a mutant animals. Gr8a was narrowly required for responding to L-canavanine, in contrast to Gr66a, which was broadly required for responding to other noxious tastants. Our data suggest that GR8a and GR66a are subunits of an L-canavanine receptor and that GR8a contributes to the specificity for L-canavanine.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
L-Canavanine, ≥98% (TLC), powder, from Canavalia ensiformis